Learn More
Research Products International Corp Methanol, ACS Grade, 4 Liter Bottle
SDP
Catalog No. 501368354
Encompass_Preferred
Colorless liquid
Flammable, Flash Point: 12°C
Boiling Point: 64-65°C
WARNING! This product can expose you to Methyl alcohol, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-136-8354 Supplier Research Products International Corp Supplier No. M220554000.0
Colorless liquid
Flammable, Flash Point: 12°C
Boiling Point: 64-65°C
WARNING! This product can expose you to Methyl alcohol, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.