Learn More
Sigma Aldrich Fine Chemicals Biosciences Methanol, 67-56-1, MFCD00004595, 4 L
Catalog No. NC1185070
Encompass
Linear Formula: CH3OH, Molecular Weight: 32.04, ACS reagent, ≥99.8%, Synonym: Methyl alcohol. Methanol has been used as a solvent for the sodium dodecyl sulfate-polyacrylamide gel electrophoretic (SDS-PAGE) analysis of alkaline protease isoforms isolated from larva of the spotted sand bass (Paralabrax maculatofasciatus). It has been used in the isolation and quantitative determination of carotenes and xanthophylls from Capsicum annum pericarp extracts by HPLC.
Catalog No. NC1185070 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1793374L
Linear Formula: CH3OH, Molecular Weight: 32.04, ACS reagent, ≥99.8%, Synonym: Methyl alcohol. Methanol has been used as a solvent for the sodium dodecyl sulfate-polyacrylamide gel electrophoretic (SDS-PAGE) analysis of alkaline protease isoforms isolated from larva of the spotted sand bass (Paralabrax maculatofasciatus). It has been used in the isolation and quantitative determination of carotenes and xanthophylls from Capsicum annum pericarp extracts by HPLC.