Learn More
Sigma Aldrich Fine Chemicals Biosciences Methanol ACS reagent 99.81L
Catalog No. 502897330
Encompass
Methanol undergoes thermochemical conversion to C2-C10 hydrocarbons in the presence of shape-selective zeolites has been reported. Its oxidation on Ru-Pt catalyst system by ruthenium ad-atoms has been proposed.
Catalog No. 50-289-7330 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1793371L
Methanol undergoes thermochemical conversion to C2-C10 hydrocarbons in the presence of shape-selective zeolites has been reported. Its oxidation on Ru-Pt catalyst system by ruthenium ad-atoms has been proposed.