Learn More
Sigma Aldrich Fine Chemicals Biosciences Methanol ACS reagent 99.8500ML
Catalog No. 502899283
Encompass
Methanol undergoes thermochemical conversion to C2-C10 hydrocarbons in the presence of shape-selective zeolites has been reported. Its oxidation on Ru-Pt catalyst system by ruthenium ad-atoms has been proposed.
Catalog No. 50-289-9283 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 179337500ML
Methanol undergoes thermochemical conversion to C2-C10 hydrocarbons in the presence of shape-selective zeolites has been reported. Its oxidation on Ru-Pt catalyst system by ruthenium ad-atoms has been proposed.