Learn More
Sigma Aldrich Fine Chemicals Biosciences Methanol ACS spectrophotom1L
Catalog No. 502898690
Encompass
Thermochemical conversion of methanol to C2-C10 hydrocarbons in the presence of shape-selective zeolites has been reported. Its oxidation on Ru-Pt catalyst system by ruthenium ad-atoms has been proposed.
Catalog No. 50-289-8690 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1549031L
Thermochemical conversion of methanol to C2-C10 hydrocarbons in the presence of shape-selective zeolites has been reported. Its oxidation on Ru-Pt catalyst system by ruthenium ad-atoms has been proposed.