Learn More
TRIBIOSCIENCE INC Methanol Assay Kit 100 Tests
SDP
Catalog No. NC3942427
Encompass_Preferred
Tribioscience Methanol Assay Kit provides a rapid, simple, reproducible, and sensitive tool for methanol quantitation in plasma, serum, urine, and other samples. The methanol assay uses coupling enzyme reactions to produce color and fluorescence. The change in color intensity of the reaction product at 570 nm or fluorescence intensity at λex/em = 530/590 nm is directly proportional to methanol concentration in the sample. The fluorometric assay is more sensitive than colorimetric assay.
Catalog No. NC3942427 Supplier TRIBIOSCIENCE INC Supplier No. TBS2018100
Tribioscience Methanol Assay Kit provides a rapid, simple, reproducible, and sensitive tool for methanol quantitation in plasma, serum, urine, and other samples. The methanol assay uses coupling enzyme reactions to produce color and fluorescence. The change in color intensity of the reaction product at 570 nm or fluorescence intensity at λex/em = 530/590 nm is directly proportional to methanol concentration in the sample. The fluorometric assay is more sensitive than colorimetric assay.