Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Jade Scientific, Inc METHANOL BIO 56L
SDP

Catalog No. 50326158 Shop All Jade Scientific, Inc Products
Encompass_Preferred

Methanol Bio 56l. 67-56-1 MFCD00004595

Catalog No. 50-326-158 Supplier Jade Scientific, Inc Supplier No. BJBC23356
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methanol Bio 56l. 67-56-1 MFCD00004595

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.