Learn More
Sigma Aldrich Fine Chemicals Biosciences Methanol BioReagent 99.931L
Catalog No. 502897853
Encompass
Methanol (MeOH) is an organic solvent with a wide range of industrial applications. It is the main component of lithium batteries. It can be prepared by hydrogenation of CO2 under various conditions. Shape-selective zeolite-assisted transformation of methanol to C2-C10 hydrocarbons has been studied. MeOH is a toxic alcohol employed for denaturing ethyl alcohol. A study reports the preparation of biodiesel from canola oil using methanol under supercritical conditions. Low water content minimal residue and clean UV spectra are the special characteristics of this solvent. It can be utilized for biotechnological processes like protein sequencing peptide and oligonucleotide synthesis.
Catalog No. 50-289-7853 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 4944371L
Methanol (MeOH) is an organic solvent with a wide range of industrial applications. It is the main component of lithium batteries. It can be prepared by hydrogenation of CO2 under various conditions. Shape-selective zeolite-assisted transformation of methanol to C2-C10 hydrocarbons has been studied. MeOH is a toxic alcohol employed for denaturing ethyl alcohol. A study reports the preparation of biodiesel from canola oil using methanol under supercritical conditions. Low water content minimal residue and clean UV spectra are the special characteristics of this solvent. It can be utilized for biotechnological processes like protein sequencing peptide and oligonucleotide synthesis.