Learn More
Medchemexpress LLC Methanol-d | 1455-13-6 | MFCD00044213 | >99.0% | 33.05 | CH3OD | 10 G
SDP
Catalog No. 5000301739
Encompass_Preferred
Methanol-d1 (CH3OD) is a deuterium-labeled form of methanol supplied as a high-purity stable isotope reagent for analytical and research use. It is intended for use as a tracer and as an internal standard in NMR, GC-MS, and LC-MS workflows. Manufacturer documentation includes purity and storage recommendations to preserve isotopic integrity.
  • High isotopic purity (≥99.0%).
  • Suitable as tracer and internal standard for NMR, GC-MS, and LC-MS analyses.
  • Provided with certificate of analysis and safety data sheet.
  • Stable under recommended storage; extended shelf life when stored at low temperature.
  • Available in laboratory-scale package sizes for research use.
Catalog No. 50-003-01739 Supplier Medchemexpress LLC Supplier No. HYW02591610G
Methanol-d1 (CH3OD) is a deuterium-labeled form of methanol supplied as a high-purity stable isotope reagent for analytical and research use. It is intended for use as a tracer and as an internal standard in NMR, GC-MS, and LC-MS workflows. Manufacturer documentation includes purity and storage recommendations to preserve isotopic integrity.
  • High isotopic purity (≥99.0%).
  • Suitable as tracer and internal standard for NMR, GC-MS, and LC-MS analyses.
  • Provided with certificate of analysis and safety data sheet.
  • Stable under recommended storage; extended shelf life when stored at low temperature.
  • Available in laboratory-scale package sizes for research use.