Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol-d, 99.5+% (Isotopic)

Catalog No. AA8957114
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8957114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA8957114 Supplier Thermo Scientific Chemicals Supplier No. 08957114

missing translation for 'validMainframeMsg'

CAS: 1455-13-6 | CH4O | 33.048 g/mol

Methanol-d is an intermediate for making other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1455-13-6
Molecular Formula CH4O
Molecular Weight (g/mol) 33.048
MDL Number MFCD00044213
InChI Key OKKJLVBELUTLKV-VMNATFBRSA-N
Synonym methanol-d, methan 2h ol, methyl alcohol-d, methanol-o-d1, ch3od, deutero methanol, ch 3 od, methanol-d1, methanol d, methan-d-ol, meod
PubChem CID 123113
IUPAC Name deuteriooxymethane
SMILES CO

Specifications

Quantity 25 g
Flash Point 11°C (54°F)
Refractive Index 1.327
Density 0.813 g/mL
Melting Point °C to 98°C
Boiling Point 64.7°C
Assay Percent Range >99.5% (Isotopic)
Beilstein 1730948
Solubility Information Completely miscible with water.
Formula Weight 33.05
Chemical Name or Material Methanol-d
Show More Show Less
Hazard Category H225-H301+H311+H331-H370
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P307+P311-P311-P312-P330-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 215-933-0
TSCA Yes
Recommended Storage Ambient

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.