Learn More
Medchemexpress LLC Methanol-d1 | 1455-13-6 | MFCD00044213 | ≥99.0% | 33.048 g/mol | CH3OD | 100 G
SDP
Catalog No. 5000299690
Encompass_Preferred
Methanol-d1 (CH3OD) is deuterium-labeled methanol intended for use as a tracer and as an internal standard in NMR, GC-MS, and LC-MS quantitative analyses. The reagent is provided with high isotopic and chemical purity and is suitable for routine analytical workflows.
  • High isotopic purity (≥99.0%) for accurate quantitative analysis.
  • Compatible with NMR, GC-MS, and LC-MS methodologies.
  • Available in multiple laboratory sizes, including 100 g.
  • Formula CH3OD enables straightforward solvent substitution in protocols.
  • Stable under recommended storage conditions with extended shelf life.
Catalog No. 50-002-99690 Supplier Medchemexpress LLC Supplier No. HYW025916100G
Methanol-d1 (CH3OD) is deuterium-labeled methanol intended for use as a tracer and as an internal standard in NMR, GC-MS, and LC-MS quantitative analyses. The reagent is provided with high isotopic and chemical purity and is suitable for routine analytical workflows.
  • High isotopic purity (≥99.0%) for accurate quantitative analysis.
  • Compatible with NMR, GC-MS, and LC-MS methodologies.
  • Available in multiple laboratory sizes, including 100 g.
  • Formula CH3OD enables straightforward solvent substitution in protocols.
  • Stable under recommended storage conditions with extended shelf life.