Learn More
Medchemexpress LLC Methanol-d1 | 1455-13-6 | MFCD00044213 | >99.0% | 33.048 g/mol | CH3OD | 25 G
SDP
Catalog No. 5000303987
Encompass_Preferred
Methanol-d1 (CH3OD) is a deuterium-labeled methanol used as a deuterated solvent, tracer, and internal standard in analytical chemistry and spectroscopic applications. It offers high isotopic purity for NMR, mass spectrometry, and tracer studies, and is supplied with safety documentation for handling and storage.
  • Deuterium-labeled methanol (CH3OD).
  • CAS number 1455-13-6.
  • High isotopic purity (≥99.0%).
  • Suitable as an NMR solvent and internal standard.
  • Useful as a tracer in isotopic labeling and analytical workflows.
  • Provided with an SDS for safe handling and transport.
Catalog No. 50-003-03987 Supplier Medchemexpress LLC Supplier No. 25GHYW025916
Methanol-d1 (CH3OD) is a deuterium-labeled methanol used as a deuterated solvent, tracer, and internal standard in analytical chemistry and spectroscopic applications. It offers high isotopic purity for NMR, mass spectrometry, and tracer studies, and is supplied with safety documentation for handling and storage.
  • Deuterium-labeled methanol (CH3OD).
  • CAS number 1455-13-6.
  • High isotopic purity (≥99.0%).
  • Suitable as an NMR solvent and internal standard.
  • Useful as a tracer in isotopic labeling and analytical workflows.
  • Provided with an SDS for safe handling and transport.