Learn More
Medchemexpress LLC Methanol-d1 | 1455-13-6 | MFCD00044213 | 99.8% | 33.05 g·mol⁻¹ | CH3DO | 5 G
SDP
Catalog No. 5000301456
Encompass_Preferred
Methanol-d1 (CH3DO) is the deuterium-labeled form of methanol used as an isotopically enriched solvent and tracer in analytical and synthetic chemistry. It provides a single deuterium substitution that aids NMR spectral interpretation and isotopic labeling studies. Typical properties include CAS 1455-13-6 and a molecular weight of 33.05 g·mol⁻¹.
  • High isotopic purity (reported 99.8%).
  • Useful as an NMR solvent and spectral reference.
  • Suitable for mass spectrometry and isotopic labeling experiments.
  • Volatile, low boiling solvent-handle in a fume hood.
  • Supplied in small research-scale packages (5 g) for laboratory use.
Catalog No. 50-003-01456 Supplier Medchemexpress LLC Supplier No. HYW0259165G
Methanol-d1 (CH3DO) is the deuterium-labeled form of methanol used as an isotopically enriched solvent and tracer in analytical and synthetic chemistry. It provides a single deuterium substitution that aids NMR spectral interpretation and isotopic labeling studies. Typical properties include CAS 1455-13-6 and a molecular weight of 33.05 g·mol⁻¹.
  • High isotopic purity (reported 99.8%).
  • Useful as an NMR solvent and spectral reference.
  • Suitable for mass spectrometry and isotopic labeling experiments.
  • Volatile, low boiling solvent-handle in a fume hood.
  • Supplied in small research-scale packages (5 g) for laboratory use.