Learn More
Cambridge Isotope Laboratories Methanol-D4 +0.03% V/V TMS | 811-98-3 | MFCD00044637 | 10 x 0.6 mL
Catalog No. NC1526417
Encompass_Preferred
Methanol-D4 +0.03% V/V TMS | Purity: 99.5% | Mol Wt: 36.07 | 811-98-3 | MFCD00044637 | 10 x 0.6 mL
Catalog No. NC1526417 Supplier Cambridge Isotope Laboratories Supplier No. DLM24TC10X0.6
Methanol-D4 +0.03% V/V TMS | Purity: 99.5% | Mol Wt: 36.07 | 811-98-3 | MFCD00044637 | 10 x 0.6 mL