Learn More
Sigma Aldrich Fine Chemicals Biosciences Methanol d4 34 10034
Catalog No. 502903739
Encompass
Methanol-d4 (CD3OD) is a deuterated NMR solvent useful in NMR-based research and analyses. Absolute infrared absorption intensities of CD3OD have been reported between 8000 and 350cm-1.
Catalog No. 50-290-3739 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 44475810X0.75ML
Methanol-d4 (CD3OD) is a deuterated NMR solvent useful in NMR-based research and analyses. Absolute infrared absorption intensities of CD3OD have been reported between 8000 and 350cm-1.