Learn More
Sigma Aldrich Fine Chemicals Biosciences Methanol-d4 99.8 atom D c
Catalog No. 502898998
Encompass
Methanol-d4 is a deuterated NMR solvent containing 0.03 % (v/v) TMS (trimethylsilane). It is useful in NMR-based research and analyses.
Catalog No. 50-289-8998 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 34380310X1ML
Methanol-d4 is a deuterated NMR solvent containing 0.03 % (v/v) TMS (trimethylsilane). It is useful in NMR-based research and analyses.