Learn More
Sigma Aldrich Fine Chemicals Biosciences Methanol-d4 99.8 atom D c10G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 61164610G
Encompass
Methanol-d4 (CD3OD) is a deuterated NMR solvent containing 0.05% (v/v) TMS (tetramethylsilane). It is useful in NMR-based research and analyses. Absolute infrared absorption intensities of CD3OD have been reported between 8000 and 350cm-1.
Catalog No. 50-289-9360