Learn More
Sigma Aldrich Fine Chemicals Biosciences Methanol-d4 99.8 atom D c10G
Catalog No. 502897377
Encompass
Methanol-d4 is a deuterated NMR solvent containing 0.03 % (v/v) TMS (trimethylsilane). It is useful in NMR-based research and analyses.
Catalog No. 50-289-7377 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 34380310G
Methanol-d4 is a deuterated NMR solvent containing 0.03 % (v/v) TMS (trimethylsilane). It is useful in NMR-based research and analyses.