Learn More
Cambridge Isotope Laboratories Methanol-D4 (d, 99.8%) +0.05% V/v Tms, 811-98-3, MFCD00044637, 10 g
Catalog No. NC9718448
Encompass_Preferred
Chemical Formula: CD3OD, MW: 36.07, 99.8%.
Catalog No. NC9718448 Supplier Cambridge Isotope Laboratories Supplier No. DLM24TB10
Add to Cart
Add to Cart
Chemical Formula: CD3OD, MW: 36.07, 99.8%.