Learn More
Cambridge Isotope Laboratories Methanol-d4 (d, 99.8%), Serum Bottle, 811-98-3, MFCD00044637, 10 g
Catalog No. NC9159564
Encompass_Preferred
Chemical Formula: CD3OD, MW: 36.07, 99.8%.
Catalog No. NC9159564 Supplier Cambridge Isotope Laboratories Supplier No. DLM24S10
Chemical Formula: CD3OD, MW: 36.07, 99.8%.