Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol - D4, deuteration degree min. 99.8 % for NMR spectroscopy MagniSolv™, MilliporeSigma™

Catalog No. M1060280010 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
10 mL
25 mL
100 mL
10 x 0.5 mL
10 x 0.75 mL
Packaging:
Glass Ampule
Glass Bottle
Septum Ampule
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1060280010 10 mL Septum Ampule
M1060280009 10 x 0.75 mL Glass Ampule
M1060280025 25 mL Glass Bottle
M1060280100 100 mL Glass Bottle
M1060280001 1 mL Glass Ampule
M1060280005 10 x 0.5 mL Glass Ampule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. M1060280010 Supplier MilliporeSigma Supplier No. 1.06028.0010
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For NMR Spectroscopy

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 811-98-3
Molecular Formula CH4O
Molecular Weight (g/mol) 36.066
InChI Key OKKJLVBELUTLKV-MZCSYVLQSA-N
Synonym methanol-d4, perdeuteromethanol, tetradeuteromethanol, methan-d3-ol-d, 2h4 methanol, methyl alcohol-d4, cd3od, methyl-d3 alcohol-d, trideuterio deuteriooxy methane, methanol-d4, 99.8 atom % d
PubChem CID 71568
IUPAC Name trideuterio(deuteriooxy)methane
SMILES CO

Specifications

Physical Form Liquid
Color Colorless
Viscosity 0.597mPa.s (20°C/68°F)
Packaging Septum Ampule
Quantity 10 mL
Odor Characteristic
Vapor Pressure 128 hPa at 20°C (68°F)
Assay Percent Range 99.8% min.
UN Number 1230
Formula Weight 36.07
Chemical Name or Material Methanol - D4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.