Learn More
Methanol-d4, for NMR, 99.8 atom % D
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Liquid |
| Color | Colorless |
| Packaging | Glass Bottle |
| Deuterium (D) | 99.75% min. |
| Density | 0.8880 g/mL |
| Water | 0.05% max. (Coulometric) |
| Refractive Index | 1.3262 |
| Quantity | 100 g |
| Flash Point | 12°C |
| Melting Point | -99.0°C |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement: Causes damage to organs. Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Highly flammable liquid and vapor. |
| Precautionary Statement | GHS P Statement: Immediately call a POISON CENTER or doctor/physician. IF exposed: Call a POISON CENTER or doctor/physician. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.