Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8 atom % D

Catalog No. AC329920250
Change view
Click to view available options
Quantity:
5 mL
10 mL
25 mL
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC329920250 25 mL Glass Bottle
AC329920050 5 mL Glass Bottle
AC329920100 10 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC329920250 Supplier Thermo Scientific Chemicals Supplier No. 329920250
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Methanol-d4, CH4O, CAS Number-811-98-3, methan-d3-ol-d, 2h4 methanol, methyl-d3 alcohol-d, perdeuteromethanol, methanol-d4, tetradeuteromethanol, trideuterio deuteriooxy methane, methanol-d4, 99.8 atom % d, methyl alcohol-d4, cd3od, 50mL, 01, III, 1187, Colorless, 0.8880g/mL, 99.75% min., 212-378-6 | CAS: 811-98-3 | CH4O | 36.066 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 811-98-3
Molecular Formula CH4O
MDL Number MFCD00044637
InChI Key OKKJLVBELUTLKV-MZCSYVLQSA-N
Synonym trideuterio deuteriooxy methane, perdeuteromethanol, methyl-d3 alcohol-d, methan-d3-ol-d, methyl alcohol-d4, tetradeuteromethanol, 2h4 methanol, cd3od, methanol-d4, methanol-d4, 99.8 atom % d
PubChem CID 71568

Specifications

Physical Form Liquid
Color Colorless
Packaging Glass Bottle
Deuterium (D) 99.75% min.
Water 0.05% max. (Coulometric)
Flash Point 12°C
Refractive Index 1.3262
Density 0.8880 g/mL
Quantity 25 mL
Melting Point -99.0°C
Boiling Point 65.0°C
Linear Formula CD3OD
Beilstein 01, III, 1187
SMILES [2H]OC([2H])([2H])[2H]
IUPAC Name (²H₃)methan(²H)ol
Molecular Weight (g/mol) 36.07
Formula Weight 36.06
Grade NMR
Chemical Name or Material Methanol-d4, 99.8 atom % D
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:

Causes damage to organs.

Toxic if swallowed.

Toxic in contact with skin.

Toxic if inhaled.

Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:

Immediately call a POISON CENTER or doctor/physician.

IF exposed: Call a POISON CENTER or doctor/physician.

IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

Ground/Bond container and receiving equipment.



WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-378-6
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.