Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol-d4,

Catalog No. AC464040250
Change view
Click to view available options
Quantity:
1 mL
10 mL
25 mL
50 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464040250 25 mL
AC464040100 1 mL
AC464040101 10 mL
AC464040500 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. AC464040250 Supplier Thermo Scientific Chemicals Supplier No. 464040250
Add to Cart
This item is not returnable. View return policy

CD4O | 36.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula CD4O
Molecular Weight (g/mol) 36.06

Specifications

Density 0.8880 g/mL
Quantity 25 mL
Flash Point 12°C
Refractive Index 1.3262
Specific Gravity 0.8880
Melting Point -99.0°C
Boiling Point 65.0°C
CAS 811-98-3
Solubility Information Solubility aq. soln.: miscible. Other solubilities: miscible with ethanol, ether, benzene, ketones and, most other organic solvents
Grade NMR
Chemical Name or Material Methanol-d4
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Causes damage to organs.

Toxic if swallowed.

Toxic in contact with skin.

Toxic if inhaled.

Highly flammable liquid and vapor.

Precautionary Statement GHS P Statement Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.

No smoking.

IF ON SKIN (or hair): Take off immediately all contaminated clothing.

Rinse skin with water or shower Wash face, hands and any exposed skin thoroughly after handling.

Wear protective gloves/protective clothing.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician.

EINECSNumber 212-378-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.