Learn More
Sigma Aldrich Fine Chemicals Biosciences Methanol suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, >=99.9% | 67-56-1 | MFCD00004595 | 4L
Catalog No. 501883491
Encompass
Methanol suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, >=99.9% | Purity: >=99.9% | Mol Wt: 32.04 | 67-56-1 | MFCD00004595 | 4L
Catalog No. 50-188-3491 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 4391934L
Methanol suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, >=99.9% | Purity: >=99.9% | Mol Wt: 32.04 | 67-56-1 | MFCD00004595 | 4L