Learn More
Sigma Aldrich Fine Chemicals Biosciences Methanol HPLC Plus 99.9
Catalog No. 502907988
Encompass
Methanol (MeOH) is an organic solvent with a wide range of industrial applications. It is the main component of lithium batteries. It can be prepared by hydrogenation of CO2 under various conditions. Shape-selective zeolite-assisted transformation of methanol to C2-C10 hydrocarbons has been studied. MeOH is a toxic alcohol employed for denaturing ethyl alcohol. A study reports the preparation of biodiesel from canola oil using methanol under supercritical conditions.
Catalog No. 50-290-7988 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 6463774X4L
Methanol (MeOH) is an organic solvent with a wide range of industrial applications. It is the main component of lithium batteries. It can be prepared by hydrogenation of CO2 under various conditions. Shape-selective zeolite-assisted transformation of methanol to C2-C10 hydrocarbons has been studied. MeOH is a toxic alcohol employed for denaturing ethyl alcohol. A study reports the preparation of biodiesel from canola oil using methanol under supercritical conditions.