Learn More
YSI 3P LifeScience Methanol Membranes, 4 each
Catalog No. NC9677419
Encompass
Methanol Membranes. Used to measure methanol on YSI 2900 Series Analyzers.
Catalog No. NC9677419 Supplier YSI 3P LifeScience Supplier No. 2725
Methanol Membranes. Used to measure methanol on YSI 2900 Series Analyzers.