Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111017869 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
3 x 1.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7869 3 x 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7869 Supplier MilliporeSigma Supelco Supplier No. PHR13723X1.5ML
Add to Cart
Add to Cart
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Methanol is a widely used polar organic solvent in chemistry, biochemistry, and molecular biology. It is a common solvent in HPLC and mass spectrometry (MS) of small molecules and of biomolecules such as oligonucleotides and proteins. Methanol is also utilized in high performance capillary electrophoresis. Industrial applications of methanol include the manufacture of formaldehyde and methyl esters of organic and inorganic acids. Methanol is used as a solvent and solvent adjuvant for polymers, and as a softening agent for pyroxylin plastics.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Vapor Pressure 410 mmHg (50°C)
Vapor Density 1.11 (vs. air)
Density 0.791 g/mL (at 25°C)
Physical Form Neat
Linear Formula CH3OH
MDL Number MFCD00004595
Melting Point -98°C
Boiling Point 64.7°C
Synonym Methanol; Methyl alcohol
Quantity 3 x 1.5 mL
Formula Weight 32.04
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.