Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Methanol, Reagent A.C.S. (Methyl Alcohol) 1 QT
SDP

Catalog No. 5027158
Encompass_Preferred

(Methyl Alcohol)
CAS #67-56-1 CH₃OH F.W. 32.04
Assay: 99.8%
Boiling point: 2°C max

Catalog No. 50-271-58 Supplier Electron Microscopy Sciences Supplier No. 18510
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Methyl Alcohol)
CAS #67-56-1 CH₃OH F.W. 32.04
Assay: 99.8%
Boiling point: 2°C max

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.