Learn More
Research Products International Corp Methanol, Scintillation Grade, 4 Liters
SDP
Catalog No. 50212981
Encompass_Preferred
Product Specifications:
  • Water: 0.20% max
  • Evaporation Residue: 0.001% max
WARNING:  This product can expose you to Methyl Alcohol, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-212-981 Supplier Research Products International Corp Supplier No. 111053
Product Specifications:
  • Water: 0.20% max
  • Evaporation Residue: 0.001% max
WARNING:  This product can expose you to Methyl Alcohol, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.