Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol Solution contains 0.10 (v/v) formic acid, UHPLC, for mass spectrometry, 99.5, ≥99.5%, MilliporeSigma™ Supelco™

Catalog No. 111017952 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
4 x 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7952 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7952 Supplier MilliporeSigma Supelco Supplier No. 6325464X4L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 67-56-1
Solution Contains 0.10 % (v/v) formic acid
Vapor Pressure 410 mmHg
Vapor Density 1.11 (vs. air)
Linear Formula CH4 O
MDL Number MFCD00004595
Melting Point -98°C
Boiling Point 65.4°C
Type Organic Standard
Quantity 4 x 4 L
Formula Weight 32.04
Percent Purity ≥99.5%
Grade UHPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.