Learn More
YSI 3P LifeScience Methanol Standards Kit, 1.0 g/L and 2.5 g/L
Catalog No. NC9683649
Encompass
1.0 g/L and 2.5 g/L Methanol Standard solutions for use with YSI 2900 Series Analyzers. Kit contains 3 bottles of 1.0 g/L standard and 2 bottles of 2.5 g/L standard.
Catalog No. NC9683649 Supplier YSI 3P LifeScience Supplier No. 2726
1.0 g/L and 2.5 g/L Methanol Standard solutions for use with YSI 2900 Series Analyzers. Kit contains 3 bottles of 1.0 g/L standard and 2 bottles of 2.5 g/L standard.