Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, Supelco

Catalog No. 11100271 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-271 4 L
11-100-257 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-271 Supplier MilliporeSigma Supelco Supplier No. 1060074000
Add to Cart
Add to Cart

Gradient grade for liquid chromatography LiChrosolv™ Reag. Ph Eur

  • High Purity: Formulated with minimal impurities, ensuring reliable and reproducible results essential for sensitive analytical applications.
  • Excellent UV Transparency: Provides optimal UV transmittance, facilitating accurate detection and enhancing sensitivity in HPLC analyses.
  • Low Evaporation Residue: Minimizes contamination risk, leading to cleaner separations and improved analytical reliability.
  • Batch-to-Batch Consistency: Ensures uniform quality and performance across different production batches, enhancing reproducibility and confidence in analytical results.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methanol
CAS 67-56-1
Quantity 4 L
Boiling Point 64.7°C (lit.)
Molecular Weight (g/mol) 32.04
Percent Purity Unknown
Grade HPLC Gradient
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.