Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide Solution, 1 mol/L (1 N), Supelco

Catalog No. 11100248 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-248 1 Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-248 Supplier MilliporeSigma Supelco Supplier No. 1.09956.0050
Only null left
Add to Cart
Add to Cart
  • Sodium hydroxide solution is used as a key reactive medium or treatment agent across diverse applications in environmental science, materials engineering, and chemical synthesis.
  • In both sets of studies, sodium hydroxide solution enhances material performance, such as improving adsorption capacity, strengthening geopolymers and stabilized soils, or optimizing functional properties of processed materials.
  • Research in both groups employs sodium hydroxide solution in aqueous systems to modify or process solid matrices, including biochar, soils, polymers, and biomaterials.
  • Across all referenced work, sodium hydroxide solution plays a critical role in developing advanced technologies and processes that support industrial, environmental, and biotechnological applications.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Hydroxide Solution
CAS 1310-73-2
Concentration 0.1 mol/L (0.1 N)
Quantity 1 Bottle
Molecular Weight (g/mol) 40.00
Percent Purity Unknown
Grade Technical
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.