Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, for UHPLC-MS LiChrosolv™, Supelco

Catalog No. 11100266 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-266 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-266 Supplier MilliporeSigma Supelco Supplier No. 1037261002
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Methanol
CAS 67-56-1
Quantity 1 L
Boiling Point 64.7°C (lit.)
Molecular Weight (g/mol) 32.04
Percent Purity Unknown
Grade Ungraded
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.