Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methanone, [1-(4-methoxyphenyl)-1H-benzimidazol-5-yl]-1-piperidinyl- | 452077-89-3 | 98.8% | 335.40 | C20H21N3O2 | 10 MG
SDP

Catalog No. 5000226532
Encompass_Preferred

A research-use small molecule inhibitor of human NAD(+)-dependent 15-hydroxyprostaglandin dehydrogenase intended for biochemical and cell-based studies. Supplied as solid and as a DMSO solution, the compound is characterized by its molecular properties and accompanied by a certificate of analysis.

  • Selective inhibitor of human NAD(+)-dependent 15-hydroxyprostaglandin dehydrogenase.
  • Suitable for enzyme inhibition assays and cell-based studies.
  • Available as solid (5, 10, 25, 50, 100 MG) and as 10 mM solution in DMSO.
  • Purity 98.8% as supplied.
  • Molecular weight 335.40; formula C20H21N3O2.

Catalog No. 50-002-26532 Supplier Medchemexpress LLC Supplier No. HY11772310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A research-use small molecule inhibitor of human NAD(+)-dependent 15-hydroxyprostaglandin dehydrogenase intended for biochemical and cell-based studies. Supplied as solid and as a DMSO solution, the compound is characterized by its molecular properties and accompanied by a certificate of analysis.

  • Selective inhibitor of human NAD(+)-dependent 15-hydroxyprostaglandin dehydrogenase.
  • Suitable for enzyme inhibition assays and cell-based studies.
  • Available as solid (5, 10, 25, 50, 100 MG) and as 10 mM solution in DMSO.
  • Purity 98.8% as supplied.
  • Molecular weight 335.40; formula C20H21N3O2.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.