Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methanone, [1-(4-methoxyphenyl)-1H-benzimidazol-5-yl]-1-piperidinyl- | 452077-89-3 | 98.8% | 335.40 | C20H21N3O2 | 100 MG
SDP

Catalog No. 5000227633
Encompass_Preferred

This small-molecule inhibitor targets human NAD(+)-dependent 15-hydroxyprostaglandin dehydrogenase (15-PGDH) and is supplied as a solid for research use only. It is intended for biochemical and cellular assays to study prostaglandin metabolism and enzyme inhibition.

  • Inhibits human NAD(+)-dependent 15-PGDH.
  • Intended for in vitro biochemical and cellular assays.
  • Supplied as a solid with high purity (98.8%).
  • Molecular weight 335.40 g/mol.
  • Available in multiple pack sizes, including 100 mg.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-27633 Supplier Medchemexpress LLC Supplier No. HY117723100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This small-molecule inhibitor targets human NAD(+)-dependent 15-hydroxyprostaglandin dehydrogenase (15-PGDH) and is supplied as a solid for research use only. It is intended for biochemical and cellular assays to study prostaglandin metabolism and enzyme inhibition.

  • Inhibits human NAD(+)-dependent 15-PGDH.
  • Intended for in vitro biochemical and cellular assays.
  • Supplied as a solid with high purity (98.8%).
  • Molecular weight 335.40 g/mol.
  • Available in multiple pack sizes, including 100 mg.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.