Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methanone, [1-(4-methoxyphenyl)-1H-benzimidazol-5-yl]-1-piperidinyl- | 452077-89-3 | 98.8% | 335.40 | C20H21N3O2 | 25 MG
SDP

Catalog No. 5000226263
Encompass_Preferred

MF-PGDH-008 is a small-molecule inhibitor of human NAD(+)-dependent 15-hydroxyprostaglandin dehydrogenase (15-PGDH) designed for in vitro research use. It is supplied as an off-white to pink solid with a molecular weight of 335.40 g/mol and high reported purity. The compound is soluble in DMSO and has defined storage recommendations for powder and solution forms.

  • Inhibits human NAD(+)-dependent 15-hydroxyprostaglandin dehydrogenase.
  • High purity (98.8%), suitable for research applications.
  • Molecular weight 335.40 g/mol, molecular formula C20H21N3O2.
  • Physical form: solid, off-white to pink, convenient for handling.
  • Solubility: readily soluble in DMSO (100 mg/mL); ultrasonic assistance recommended.
  • Storage: recommended conditions given for powder and in-solution stability.
  • Suitable for in vitro biochemical and cell-based assays.

Catalog No. 50-002-26263 Supplier Medchemexpress LLC Supplier No. HY11772325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MF-PGDH-008 is a small-molecule inhibitor of human NAD(+)-dependent 15-hydroxyprostaglandin dehydrogenase (15-PGDH) designed for in vitro research use. It is supplied as an off-white to pink solid with a molecular weight of 335.40 g/mol and high reported purity. The compound is soluble in DMSO and has defined storage recommendations for powder and solution forms.

  • Inhibits human NAD(+)-dependent 15-hydroxyprostaglandin dehydrogenase.
  • High purity (98.8%), suitable for research applications.
  • Molecular weight 335.40 g/mol, molecular formula C20H21N3O2.
  • Physical form: solid, off-white to pink, convenient for handling.
  • Solubility: readily soluble in DMSO (100 mg/mL); ultrasonic assistance recommended.
  • Storage: recommended conditions given for powder and in-solution stability.
  • Suitable for in vitro biochemical and cell-based assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.