Learn More
Medchemexpress LLC Methanone, [1-(4-methoxyphenyl)-1H-benzimidazol-5-yl]-1-piperidinyl- | 452077-89-3 | 98.8% | 335.40 | C20H21N3O2 | 5 MG
SDP
Catalog No. 5000227200
Encompass_Preferred
A research-grade small-molecule inhibitor of human NAD(+)-dependent 15-hydroxyprostaglandin dehydrogenase (15-PGDH), provided for laboratory research use. The compound is supplied as an off-white to pink solid and is also available as a 10 mM solution in DMSO; common pack sizes include 5 mg.
  • Research-grade inhibitor of 15-PGDH for in vitro studies.
  • Available as a solid and as a 10 mM, 1 mL solution in DMSO.
  • Common pack sizes include 5 mg for small-scale experiments.
  • High purity (98.8%) with Certificate of Analysis.
  • Molecular formula C20H21N3O2; molecular weight 335.40.
  • For research use only; not for human or clinical applications.
Catalog No. 50-002-27200 Supplier Medchemexpress LLC Supplier No. HY1177235MG
A research-grade small-molecule inhibitor of human NAD(+)-dependent 15-hydroxyprostaglandin dehydrogenase (15-PGDH), provided for laboratory research use. The compound is supplied as an off-white to pink solid and is also available as a 10 mM solution in DMSO; common pack sizes include 5 mg.
  • Research-grade inhibitor of 15-PGDH for in vitro studies.
  • Available as a solid and as a 10 mM, 1 mL solution in DMSO.
  • Common pack sizes include 5 mg for small-scale experiments.
  • High purity (98.8%) with Certificate of Analysis.
  • Molecular formula C20H21N3O2; molecular weight 335.40.
  • For research use only; not for human or clinical applications.