Learn More
Medchemexpress LLC Methanone, [1-(4-methoxyphenyl)-1H-benzimidazol-5-yl]-1-piperidinyl- | 452077-89-3 | 98.8% | 335.40 | C20H21N3O2 | 50 MG
SDP
Catalog No. 5000228219
Encompass_Preferred
MF-PGDH-008 is a small-molecule inhibitor of human NAD(+)-dependent 15-hydroxyprostaglandin dehydrogenase provided for research use. It is characterized for biochemical and preclinical studies and supplied at high purity with detailed storage and solubility information.
  • Inhibits human 15-hydroxyprostaglandin dehydrogenase in biochemical assays.
  • High purity suitable for research use (98.8%).
  • Documented solubility: DMSO 100 mg/mL; compatible with common in vivo formulations.
  • Recommended storage: powder at -20°C; solvent solutions at -80°C for long-term stability.
  • Includes CAS identifier 452077-89-3 for unambiguous compound identification.
Catalog No. 50-002-28219 Supplier Medchemexpress LLC Supplier No. HY11772350MG
MF-PGDH-008 is a small-molecule inhibitor of human NAD(+)-dependent 15-hydroxyprostaglandin dehydrogenase provided for research use. It is characterized for biochemical and preclinical studies and supplied at high purity with detailed storage and solubility information.
  • Inhibits human 15-hydroxyprostaglandin dehydrogenase in biochemical assays.
  • High purity suitable for research use (98.8%).
  • Documented solubility: DMSO 100 mg/mL; compatible with common in vivo formulations.
  • Recommended storage: powder at -20°C; solvent solutions at -80°C for long-term stability.
  • Includes CAS identifier 452077-89-3 for unambiguous compound identification.