Learn More
Medchemexpress LLC Methanone, [2-[(3,5-dimethyl-1,2-oxazol-4-yl)methylsulfanyl]phenyl]-[4-(3-CF3-phenyl)piperazin-1-yl] | 1182129-77-6 | MFCD15344345 | 98.7% | 475.53 | C24H24F3N3O2S | 100MG
SDP
Catalog No. 5000237448
Encompass_Preferred
RU-302 is a small-molecule pan-TAM inhibitor that disrupts the interface between the TAM Ig1 ectodomain and the Gas6 LG domain, inhibiting Gas6-inducible Axl receptor activation in cell assays (low micromolar IC50) and demonstrating antitumor activity in lung cancer models. The compound is supplied as a research reagent for biochemical and cell-based studies.
  • Pan-TAM inhibitor that targets Axl receptor activation.
  • Shown to inhibit Gas6-inducible Axl signaling with low micromolar potency in cell assays.
  • Demonstrated suppression of lung cancer tumor growth in preclinical studies.
  • High purity (98.7%) suitable for biochemical and cell-based assays.
  • Molecular formula C24H24F3N3O2S; molecular weight 475.53.
  • Solid, white to off-white; soluble in DMSO.
  • Available in multiple pack sizes for laboratory use.
Catalog No. 50-002-37448 Supplier Medchemexpress LLC Supplier No. HY124066100MG
RU-302 is a small-molecule pan-TAM inhibitor that disrupts the interface between the TAM Ig1 ectodomain and the Gas6 LG domain, inhibiting Gas6-inducible Axl receptor activation in cell assays (low micromolar IC50) and demonstrating antitumor activity in lung cancer models. The compound is supplied as a research reagent for biochemical and cell-based studies.
  • Pan-TAM inhibitor that targets Axl receptor activation.
  • Shown to inhibit Gas6-inducible Axl signaling with low micromolar potency in cell assays.
  • Demonstrated suppression of lung cancer tumor growth in preclinical studies.
  • High purity (98.7%) suitable for biochemical and cell-based assays.
  • Molecular formula C24H24F3N3O2S; molecular weight 475.53.
  • Solid, white to off-white; soluble in DMSO.
  • Available in multiple pack sizes for laboratory use.