Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methanone, (2-amino-5-bromophenyl)phenyl- | 39859-36-4 | MFCD00059962 | 99.8% | 276.13 | C13H10BrNO | 100 G
SDP

Catalog No. 5000208532
Encompass_Preferred

2-Amino-5-bromobenzophenone is a brominated benzophenone derivative used as a synthetic intermediate in pharmaceutical and medicinal chemistry research. It is intended for research use and commonly used in chemical synthesis and compound development.

  • CAS number 39859-36-4.
  • Molecular formula C13H10BrNO.
  • Molecular weight 276.13 g/mol.
  • Purity 99.8%.
  • Appearance: light yellow solid.
  • Available in 100 G packaging.

Catalog No. 50-002-08532 Supplier Medchemexpress LLC Supplier No. HYW087426100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Amino-5-bromobenzophenone is a brominated benzophenone derivative used as a synthetic intermediate in pharmaceutical and medicinal chemistry research. It is intended for research use and commonly used in chemical synthesis and compound development.

  • CAS number 39859-36-4.
  • Molecular formula C13H10BrNO.
  • Molecular weight 276.13 g/mol.
  • Purity 99.8%.
  • Appearance: light yellow solid.
  • Available in 100 G packaging.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.