Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methanone, (2-amino-5-bromophenyl)phenyl-. | 39859-36-4 | MFCD00059962 | 99.8% | 276.13 g·mol⁻¹ | C13H10BrNO | 10 G
SDP

Catalog No. 5000208397
Encompass_Preferred

2-Amino-5-bromobenzophenone is a brominated benzophenone derivative used as an intermediate in pharmaceutical and organic synthesis. It is supplied in laboratory-scale quantities and is characterized by a high reported purity.

  • High reported purity (99.82%).
  • Molecular formula C13H10BrNO, molecular weight 276.13 g·mol⁻¹.
  • Available in multiple package sizes: 1 g, 5 g, 10 g, 25 g, 50 g, 100 g, >100 g.
  • Synonym: 2-benzoyl-4-bromoaniline.

Catalog No. 50-002-08397 Supplier Medchemexpress LLC Supplier No. HYW08742610G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Amino-5-bromobenzophenone is a brominated benzophenone derivative used as an intermediate in pharmaceutical and organic synthesis. It is supplied in laboratory-scale quantities and is characterized by a high reported purity.

  • High reported purity (99.82%).
  • Molecular formula C13H10BrNO, molecular weight 276.13 g·mol⁻¹.
  • Available in multiple package sizes: 1 g, 5 g, 10 g, 25 g, 50 g, 100 g, >100 g.
  • Synonym: 2-benzoyl-4-bromoaniline.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.