Learn More
Medchemexpress LLC Methanone, (2-chloro-10H-phenothiazin-10-yl)(4-methoxyphenyl) | 105925-39-1 | MFCD01245092 | 99.0% | 367.85 g/mol | C20H14ClNO2S | 5 MG
SDP
Catalog No. 5000223392
Encompass_Preferred
Tubulin inhibitor 6 (CAS 105925-39-1) is a small-molecule research compound used to probe microtubule dynamics and anti-mitotic mechanisms in vitro. It inhibits tubulin polymerization and demonstrates cytotoxic activity in cell-based assays, supplied in small milligram quantities for laboratory research.
  • Inhibits tubulin polymerization with an IC50 of 0.87 μM.
  • Inhibits K562 cell growth with an IC50 of 840 nM.
  • High stated purity (~99%) appropriate for biochemical and cellular assays.
  • Molecular weight 367.85 g/mol; molecular formula C20H14ClNO2S.
  • Provided in milligram-scale quantities for research use.
Catalog No. 50-002-23392 Supplier Medchemexpress LLC Supplier No. HY1361215MG
Tubulin inhibitor 6 (CAS 105925-39-1) is a small-molecule research compound used to probe microtubule dynamics and anti-mitotic mechanisms in vitro. It inhibits tubulin polymerization and demonstrates cytotoxic activity in cell-based assays, supplied in small milligram quantities for laboratory research.
  • Inhibits tubulin polymerization with an IC50 of 0.87 μM.
  • Inhibits K562 cell growth with an IC50 of 840 nM.
  • High stated purity (~99%) appropriate for biochemical and cellular assays.
  • Molecular weight 367.85 g/mol; molecular formula C20H14ClNO2S.
  • Provided in milligram-scale quantities for research use.