Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methanone, [2-hydroxy-4-(4-morpholinyl)phenyl]phenyl- | 404009-46-7 | MFCD09037528 | 99.5% | 283.32 g/mol | C17H17NO3 | 1 ML
SDP

Catalog No. 5000214447
Encompass_Preferred

AMA-37 is an arylmorpholine ATP-competitive inhibitor of DNA-dependent protein kinase (DNA-PK) used for biochemical and cellular research. The compound is provided in both solution and solid formats to support assay work and synthesis.

  • Inhibits DNA-PK with IC50 0.27 μM and shows activity against p110α (32 μM), p110β (3.7 μM), and p110γ (22 μM).
  • Available as a 10 mM solution in DMSO (1 mL) and in solid vials for scale flexibility.
  • CAS number 404009-46-7 for unambiguous chemical identification.
  • High reported purity (~99.5%) suitable for research applications.
  • Molecular weight 283.32 g/mol for formulation and dosing calculations.
  • Appropriate for DNA-PK signaling studies, kinase selectivity profiling, and preclinical assay development.

Catalog No. 50-002-14447 Supplier Medchemexpress LLC Supplier No. HY10070610MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AMA-37 is an arylmorpholine ATP-competitive inhibitor of DNA-dependent protein kinase (DNA-PK) used for biochemical and cellular research. The compound is provided in both solution and solid formats to support assay work and synthesis.

  • Inhibits DNA-PK with IC50 0.27 μM and shows activity against p110α (32 μM), p110β (3.7 μM), and p110γ (22 μM).
  • Available as a 10 mM solution in DMSO (1 mL) and in solid vials for scale flexibility.
  • CAS number 404009-46-7 for unambiguous chemical identification.
  • High reported purity (~99.5%) suitable for research applications.
  • Molecular weight 283.32 g/mol for formulation and dosing calculations.
  • Appropriate for DNA-PK signaling studies, kinase selectivity profiling, and preclinical assay development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.