Learn More
Medchemexpress LLC Methanone, (2-oxido-1,2,5-oxadiazole-3,4-diyl)bis[(2-methoxyphenyl)- (9CI) | 886536-76-1 | 99.1% | 354.31 | 25 MG
SDP
Catalog No. 5000394875
Encompass_Preferred
NCGC00351170 is an antiplatelet agent that disrupts the calcium and integrin-binding protein 1 (CIB1)-αIIbβ3 interaction. It inhibits thrombin-induced human platelet aggregation.
  • Antiplatelet agent
  • Disrupts CIB1-αIIbβ3 interaction
  • Inhibits thrombin-induced human platelet aggregation
  • For research use only
Catalog No. 50-003-94875 Supplier Medchemexpress LLC Supplier No. 25MGHY162558
NCGC00351170 is an antiplatelet agent that disrupts the calcium and integrin-binding protein 1 (CIB1)-αIIbβ3 interaction. It inhibits thrombin-induced human platelet aggregation.
  • Antiplatelet agent
  • Disrupts CIB1-αIIbβ3 interaction
  • Inhibits thrombin-induced human platelet aggregation
  • For research use only