Learn More
Medchemexpress LLC Methanone, [(2S,3aS,7aS)-octahydro-1-[[(1R,2R)-2-phenylcyclopropyl]carbonyl]-1H-indol-2-yl]-3-thiazolidinyl- | 176797-26-5 | C22H28N2O2S | 50 MG
SDP
Catalog No. 5000375815
Encompass_Preferred
S 17092 (S 17092-1) is an orally active cerebral prolyl-endopeptidase (PEP) inhibitor with an IC50 of 1.2 nM. It inhibits cell apoptosis by slowing down the degradation of neuroactive peptides, thereby enhancing memory function. It is suitable for research on memory impairment and cognitive disorders associated with cerebral aging.
  • Orally active cerebral prolyl-endopeptidase (PEP) inhibitor
  • Inhibits cell apoptosis
  • Enhances memory function
  • Useful for research on memory impairment and cognitive disorders
  • Long-acting PEP inhibitor with a half-life of over 9 hours
Catalog No. 50-003-75815 Supplier Medchemexpress LLC Supplier No. HY1511450MG
S 17092 (S 17092-1) is an orally active cerebral prolyl-endopeptidase (PEP) inhibitor with an IC50 of 1.2 nM. It inhibits cell apoptosis by slowing down the degradation of neuroactive peptides, thereby enhancing memory function. It is suitable for research on memory impairment and cognitive disorders associated with cerebral aging.
  • Orally active cerebral prolyl-endopeptidase (PEP) inhibitor
  • Inhibits cell apoptosis
  • Enhances memory function
  • Useful for research on memory impairment and cognitive disorders
  • Long-acting PEP inhibitor with a half-life of over 9 hours