Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methanone, [3-[[4-(aminomethyl)-6-(trifluoromethyl)-2-pyridinyl]oxy]phenyl][(3R,4R)-3-fluoro-4-hydro | 2007885-39-2 | 99.5% | 399.34 g/mol | C18H17F4N3O3 | 250 MG
SDP

Catalog No. 5000239734
Encompass_Preferred

Lenumlostat is a small-molecule inhibitor of lysyl oxidase-like 2 (LOXL2) supplied for research use in preclinical studies of fibrotic disease. The compound is characterized by analytical data and supplied as a solid for laboratory use.

  • Small-molecule LOXL2 inhibitor for research applications.
  • High purity (99.5% by LCMS).
  • Molecular weight 399.34 g/mol.
  • CAS number 2007885-39-2.
  • Available as a 250 mg solid.
  • Used in preclinical fibrosis and LOXL2 pathway studies.

Catalog No. 50-002-39734 Supplier Medchemexpress LLC Supplier No. HY107422250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lenumlostat is a small-molecule inhibitor of lysyl oxidase-like 2 (LOXL2) supplied for research use in preclinical studies of fibrotic disease. The compound is characterized by analytical data and supplied as a solid for laboratory use.

  • Small-molecule LOXL2 inhibitor for research applications.
  • High purity (99.5% by LCMS).
  • Molecular weight 399.34 g/mol.
  • CAS number 2007885-39-2.
  • Available as a 250 mg solid.
  • Used in preclinical fibrosis and LOXL2 pathway studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.