Learn More
Medchemexpress LLC Methanone, bis(4-aminophenyl)- | 611-98-3 | MFCD00038138 | 99.8% | 212.25 g/mol | C13H12N2O | 5 G
SDP
Catalog No. 5000302810
Encompass_Preferred
4,4′-Diaminobenzophenone is an aromatic diamine used as a synthetic building block and intermediate in the production of dyes, pigments, and photosensitive materials. Supplied for research use, the material is accompanied by analytical and safety documentation to support quality control and handling.
  • High purity suitable for synthetic and analytical applications.
  • Aromatic diamine functional group enables diverse chemical transformations.
  • Provided with analytical documentation (COA, H NMR, RP-HPLC) for verification.
  • Solid, off-white to yellow appearance for straightforward handling and storage.
  • Available in lab-scale packaging sizes for research workflows.
Catalog No. 50-003-02810 Supplier Medchemexpress LLC Supplier No. HYW0142905G
Add to Cart
Add to Cart
4,4′-Diaminobenzophenone is an aromatic diamine used as a synthetic building block and intermediate in the production of dyes, pigments, and photosensitive materials. Supplied for research use, the material is accompanied by analytical and safety documentation to support quality control and handling.
  • High purity suitable for synthetic and analytical applications.
  • Aromatic diamine functional group enables diverse chemical transformations.
  • Provided with analytical documentation (COA, H NMR, RP-HPLC) for verification.
  • Solid, off-white to yellow appearance for straightforward handling and storage.
  • Available in lab-scale packaging sizes for research workflows.