Learn More
AdipoGen Methansulfonic anhydride
SDP
Catalog No. 502560451
Encompass_Preferred
Chemical. CAS 03.01.7143. Formula C2H6O5S2. MW 174.2. Methanesulfonic anhydride is a building block for organic synthesis. It is used to prepare methylsulfonyl esters, also called as mesylates. It finds application in the reaction of aromatic alkylation. It reacts with dimethylsulfoxide to get sulfoxonium complex, which is a useful oxidizing agent. It is a more powerful sulfonylating reagent than the sulfonyl chloride. Used in the synthesis of R-Amino Carnitine, an inhibitor of fatty acid oxidation.
Catalog No. 50-256-0451 Supplier AdipoGen Supplier No. CDXM0698G010
Chemical. CAS 03.01.7143. Formula C2H6O5S2. MW 174.2. Methanesulfonic anhydride is a building block for organic synthesis. It is used to prepare methylsulfonyl esters, also called as mesylates. It finds application in the reaction of aromatic alkylation. It reacts with dimethylsulfoxide to get sulfoxonium complex, which is a useful oxidizing agent. It is a more powerful sulfonylating reagent than the sulfonyl chloride. Used in the synthesis of R-Amino Carnitine, an inhibitor of fatty acid oxidation.